missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP22A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572645
This item is not returnable.
View return policy
Description
DUSP22A Polyclonal Antibody for Western Blot
Activates the Jnk signaling pathway. Dephosphorylates and deactivates p38 and stress-activated protein kinase/c-Jun N-terminal kinase (SAPK/JNK).
Specifications
| DUSP22A | |
| Polyclonal | |
| Unconjugated | |
| dusp22a | |
| dual specificity phosphatase 22; dual specificity phosphatase 22a; dual specificity protein phosphatase 22-A; dusp22; dusp22a; hypothetical protein LOC436787; si:dkey-190g1.1; wu:fa11e09; zgc:92816 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 436787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q1LWL2 | |
| dusp22a | |
| KLH conjugated synthetic peptide between 172-195 amino acids from the C-terminal region of DANRE dusp22a. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction