missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16239
This item is not returnable.
View return policy
Description
DUSP22 Polyclonal specifically detects DUSP22 in Human samples. It is validated for Western Blot.
Specifications
| DUSP22 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| dual specificity phosphatase 22, dual specificity protein phosphatase 22, EC 3.1.3.16, EC 3.1.3.48, FLJ35864, homolog of mouse dual specificity phosphatase LMW-DSP2, JKAP, JNK-stimulating phosphatase 1, JSP-1, JSP1JNK-stimulatory phosphatase-1, LMWDSP2, LMW-DSP2, Low molecular weight dual specificity phosphatase 2, Mitogen-activated protein kinase phosphatase x, MKP-x, MKPXMAP kinase phosphatase x, VHX | |
| Rabbit | |
| 21 kDa | |
| 0.1 mL | |
| GPCR, MAP Kinase Signaling, Protein Phosphatase | |
| 56940 | |
| Human | |
| IgG |
| Western Blot | |
| 0.83mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NRW4 | |
| DUSP22 | |
| Recombinant protein encompassing a sequence within the center region of human DUSP22. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (93%), Rat (91%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction