missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549858
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DUSP19 protein.
Dual-specificity phosphatases.
Specifications
| DUSP19 | |
| Polyclonal | |
| Unconjugated | |
| DUSP19 | |
| 5930436K22Rik; C79103; dual specificity phosphatase 19; dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; DUSP17; DUSP19; LMWDSP3; LMW-DSP3; low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; SKRP1; Stress-activated protein kinase pathway-regulating phosphatase 1; TS-DSP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 142679 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WTR2 | |
| DUSP19 | |
| Synthesized peptide derived from internal of human DUS19 (aa 111-160). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction