missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP16, Mouse anti-Human, Clone: 3F3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00080824-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: TESSHFYSASAIYGGSASYSAYSCSQLPTCGDQVYSVRRRQKPSDRADSRRSWHEESPFEKQFKRRSCQMEFGESIMSENRSREELGKVGSQSSFSGSMEIIEVSSpecifications
| DUSP16 | |
| Monoclonal | |
| Unconjugated | |
| dual specificity phosphatase 16 | |
| KIAA1700/MGC129701/MGC129702/MKP-7/MKP7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 3F3 | |
| In 1x PBS, pH 7.4 | |
| NM_030640 | |
| DUSP16 | |
| DUSP16 (NP_085143.1, 561 a.a. ∼ 665 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 80824 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction