missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92694-0.1ml
This item is not returnable.
View return policy
Description
DUSP16 Polyclonal antibody specifically detects DUSP16 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DUSP16 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| dual specificity phosphatase 16, EC 3.1.3.16, EC 3.1.3.48, FLJ36298, FLJ40991, KIAA1700MGC129701, MAP kinase phosphatase 7, MAPK phosphatase-7, Mitogen-activated protein kinase phosphatase 7, MKP7dual specificity protein phosphatase 16, MKP-7MGC129702 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 416-665 of human DUSP16 (NP_085143.1). SLHGFSSSEDALEYYKPSTTLDGTNKLCQFSPVQELSEQTPETSPDKEEASIPKKLQTARPSDSQSKRLHSVRTSSSGTAQRSLLSPLHRSGSVEDNYHTSFLFGLSTSQQHLTKSAGLGLKGWHSDILAPQTSTPSLTSSWYFATESSHFYSASAIYGGSASYSAYSCSQLPTCGDQVYSVRRRQKPSDRADSRRSWHEESPFEKQFKRRSCQMEFGESIMSENRSREELGKVGSQSSFSGSMEIIEVS | |
| 0.1 mL | |
| Protein Phosphatase, Signal Transduction | |
| 80824 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction