missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92313-0.1ml
This item is not returnable.
View return policy
Description
DUSP15 Polyclonal antibody specifically detects DUSP15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DUSP15 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| dual specificity phosphatase 15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-90 of human DUSP15 (NP_542178.2). PGLYLGNFIDAKDLDQLGRNKITHIISIHESPQPLLQDITYLRIPVADTPEVPIKKHFKECINFIHCCRLNGGNCLVHCFA | |
| 0.1 mL | |
| Protein Phosphatase | |
| 128853 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction