missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103288
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DUSP14.
In addition to antigen recognition by the T-cell receptor, T-cell activation requires a second signal from a costimulatory receptor, such as CD28 (MIM 186760), which interacts with B7-1 (CD80; MIM 112203) and B7-2 (CD86; MIM 601020) ligands on antigen-presenting cells. CD28 costimulation induces transcription of interleukin-2 (IL2; MIM 147680) and stabilizes newly synthesized IL2 through the activation of mitogen-activated protein kinases (MAPKs), such as ERK (e.g., MAP2K4; MIM 601335) and JNK (see MIM 601158), and the subsequent creation of AP1 transcription factor (see MIM 165160). DUSP14 is a negative regulator of CD28 signaling.
Specifications
| DUSP14 | |
| Polyclonal | |
| Unconjugated | |
| DUSP14 | |
| 1110014C10Rik; 2310042C07Rik; D11Ertd395e; dual specificity phosphatase 14; dual specificity protein phosphatase 14; DUSP14; MAP kinase phosphatase 6; MAP kinase phsophatase 6; mitogen-activated protein kinase phosphatase 6; Mkp-1 like protein tyrosine phosphatase; MKP-1-like protein tyrosine phosphatase; MKP6; MKP-6; MKP-L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11072, 360580, 56405 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95147, Q9JLY7 | |
| DUSP14 | |
| A synthesized peptide derived from human DUSP14(Accession O95147), corresponding to amino acid residues A42-T92. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction