missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99994-50ul
This item is not returnable.
View return policy
Description
DUSP14 Polyclonal antibody specifically detects DUSP14 in Human samples. It is validated for ELISA
Specifications
| DUSP14 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| dual specificity phosphatase 14, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 6, Mitogen-activated protein kinase phosphatase 6, MKP-1 like protein tyrosine phosphatase, MKP-1-like protein tyrosine phosphatase, MKP-6, MKP6MKP-Ldual specificity protein phosphatase 14 | |
| Produced in rabbits immunized with purified, recombinant Human DUSP14 (Accession#: O95147; Met1-His191) | |
| 50 μg | |
| Protein Phosphatase | |
| 11072 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction