missing translation for 'onlineSavingsMsg'
Learn More

DUSP14 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NB100-887

Product Code. 18283400

  • £329.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DUSP14 Polyclonal specifically detects DUSP14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DUSP14
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
dual specificity phosphatase 14, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 6, Mitogen-activated protein kinase phosphatase 6, MKP-1 like protein tyrosine phosphatase, MKP-1-like protein tyrosine phosphatase, MKP-6, MKP6MKP-Ldual specificity protein phosphatase 14
Goat
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 1 ug/ml, Immunohistochemistry 3.5 ug/ml, Immunohistochemistry-Paraffin 3.5 ug/ml, Peptide ELISA Detection limit 1:32000
NP_008957.
DUSP14
Peptide with sequence CEKESRHLMPYWGI corresponding to C-Terminus according to NP_008957.1.
0.1 mg
Protein Phosphatase
11072
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only