missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DUSP14 Polyclonal antibody specifically detects DUSP14 in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human DUSP14 (Accession#: O95147; Met1-His191) |
| Quantity | 0.1 mL |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?