missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598501
This item is not returnable.
View return policy
Description
DUSP13 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.
Specifications
| DUSP13 | |
| Polyclonal | |
| Unconjugated | |
| DUSP13 | |
| BEDP; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; DUSP13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; Mdsp; muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; TMDP; TS-DSP6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51207 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6B8I1, Q9UII6 | |
| DUSP13, DUSP13B | |
| Recombinant Human Dual specificity protein phosphatase 13 isoform A protein (1-198AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion