missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP13 Antibody [DyLight 650], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-46619C
This item is not returnable.
View return policy
Description
DUSP13 Polyclonal antibody specifically detects DUSP13 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| DUSP13 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| DyLight 650 | |
| Branching-enzyme interacting DSP, dual specificity phosphatase 13, Dual specificity phosphatase SKRP4, dual specificity protein phosphatase 13, dual specificity protein phosphatase 13 isoform MDSP, dual-specificity phosphatase SKRP4, DUSP13A, DUSP13B, EC 3.1.3.16, EC 3.1.3.48, FLJ32450, MDSPbranching-enzyme interacting dual-specificity protein phosphatase, Muscle-restricted DSP, Testis- and skeletal-muscle-specific DSP, TMDPBEDPSKRP4 | |
| dual specificity phosphatase 13. (Uniprot# Q9UII6) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51207 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction