missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31197
This item is not returnable.
View return policy
Description
DUSP12 Polyclonal specifically detects DUSP12 in Human samples. It is validated for Western Blot.
Specifications
| DUSP12 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DUSP12 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human DUSP12. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| dual specificity phosphatase 12, dual specificity protein phosphatase 12, Dual specificity tyrosine phosphatase YVH1, DUSP1YVH1 protein-tyrosine phosphatase ortholog, EC 3.1.3.48, serine/threonine specific protein phosphatase, TDSP4, YVH1EC 3.1.3.16 | |
| Rabbit | |
| 38 kDa | |
| 100 μL | |
| Protein Phosphatase | |
| 11266 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction