missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68919
This item is not returnable.
View return policy
Description
DUSP11 Polyclonal specifically detects DUSP11 in Mouse samples. It is validated for Western Blot.
Specifications
| DUSP11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dual specificity phosphatase 11 (RNA/RNP complex 1-interacting), Dual specificity protein phosphatase 11, EC 3.1.3.-, Phosphatase that interacts with RNA/RNP complex 1, PIR1RNA/RNP complex-interacting phosphatase, RNA/RNP complex-1-interacting phosphatase, serine/threonine specific protein phosphatase | |
| Rabbit | |
| 38 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6NXK5 | |
| DUSP11 | |
| Synthetic peptides corresponding to Dusp11 (dual specificity phosphatase 11 (RNA/RNP complex 1-interacting)) The peptide sequence was selected from the N terminal of Dusp11. Peptide sequence GQRMPGTRFIAFKVPLQKKFEAKLMPEECFSPLDLFNKIQEQNEELGLII The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 8446 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction