missing translation for 'onlineSavingsMsg'
Learn More

DUSP10 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18406932 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406932 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406932 Supplier Novus Biologicals Supplier No. NBP130062G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DUSP10 Polyclonal antibody specifically detects DUSP10 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP10
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 10, dual specificity phosphatase MKP-5, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 5, Mitogen-activated protein kinase phosphatase 5, MKP-5dual specificity protein phosphatase 10, MKP5serine/threonine specific protein phosphatase
Host Species Rabbit
Immunogen maps to a region between residue 432 and 482 of human dual specificity phosphatase 10 using the numbering given in entry NP_009138.1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11221
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.