missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUS3L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584569
This item is not returnable.
View return policy
Description
Immunogen sequence: DIRGKLYLAP LTTCGNLPFR RICKRFGADV TCGEMAVCTN LLQGQMSEWA LLKRHQCEDI FGVQLEGAFP DTMTKCAELL Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 96%.
DUS3L gene ontology annotations related to this gene include flavin adenine dinucleotide binding; metal ion binding; tRNA dihydrouridine synthase activity.
Specifications
| DUS3L | |
| Polyclonal | |
| Unconjugated | |
| DUS3L | |
| AI662135; AW557805; dihydrouridine synthase 3 like; dihydrouridine synthase 3-like; dihydrouridine synthase 3-like (S. cerevisiae); DUS3; DUS3L; putative zinc finger protein MA23; tRNA-dihydrouridine synthase 3-like; tRNA-dihydrouridine(47) synthase [NAD(P)(+)]-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96G46 | |
| DUS3L | |
| Recombinant protein corresponding to Human DUS3L. Recombinant protein control fragment (Product #RP-107198). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction