missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DUS2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30263
This item is not returnable.
View return policy
Description
DUS2L Polyclonal antibody specifically detects DUS2L in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DUS2L | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54920 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| dihydrouridine synthase 2-like, SMM1 homolog (S. cerevisiae) | |
| Between 443 and 493 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction