missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 3 μm Pore Size (ME 29) Mixed Cellulose Ester Membranes, Plain
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 29
- Filter Type: Plain
- Pore Size: 3 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Bubble Point: 1.3 bar
- Flow Rate: 1.2 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 6.7 sec./100 mL (at Δp = 3 mbar)
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | MCE |
| Material (Membrane) | ME 29 |
| Pore Size | 3 μm |
| Sterility | Non-sterile |
| Flow Rate | 1.2 sec./100 mL/12.5 cm² (at Δp = 0.9 bar) |
| For Use With (Application) | Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays |
| Gas Flow Rate | 6.7 sec./100 mL (at Δp = 3 mbar) |
| Surface Type | Plain |
| Diameter (English) | 1.85 in. |
| Show More |
Safety and Handling
Signal Word
- ||Warning
- Flammable solid Category 1
- Flammable solid Category 2
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contain: nitrocellulose
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?