missing translation for 'onlineSavingsMsg'
Learn More

SCAT DURAN™ YOUTILITY Laboratory Bottle

Product Code. 17734884 Shop All SCAT Europe Products
Click to view available options
Color:
Brown
Clear
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17734884

Brand: SCAT 501131

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratory bottles made of borosilicate glass for laboratories with the highest quality requirements in terms of resistance, thermal behavior and long service life.

  • High resistance to chemical influences
  • Excellent thermal behavior for use up to 140°C
  • Long service life thanks to high quality workmanship
  • Fully autoclavable at 121 or 134°C
  • In various thread sizes, shapes and colors
  • Optionally with ground neck or with wide neck opening
  • Variants with protective coating available
  • Includes blue PP screw cap

Specifications

Capacity (Metric) 1000 mL
Color Clear
Description DURAN Laboratory bottle
Material Borosilicate Glass (3.3)
Product Line Youtility
Thread Size (Metric) GL45
Shape Round
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.