Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottle Heads, with Filter Disc
Description
- Acc. to DIN 12596, with cone
- For bottles acc. to DIN 12463
- Available in a range of porosities
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Material | Duran Tubing |
| Type | Gas Washing Bottle Head with Filter Disc |
| Includes | Filter disc |
| Porosity | 0 |
| Standard Taper Joint | NS 29/32 |
| Certifications/Compliance | DIN 12596, DIN 12463 |
| Quantity | 1 Pack |