Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
Brand:  DWK Life Sciences 248449109
Code : NEW
Additional Details : Weight : 0.17000kg
 View more versions of this product
Product Code. 15292009
  • £226.00 / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
  • Tight closure due to standard ground joint
  • With DIN thread
Specifications
Specifications
10/Pk.
Tube with screw threads
GL 32, with Cone NS 29/32