Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
Brand:  DWK Life Sciences 286038407
Additional Details : Weight : 12.50000kg
 View more versions of this product
Product Code. 15824528
  • £407.00 / Pack of 50
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
  • With groove, polished
  • The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
  • Solid glass tap plugs
  • Taper 1:10
Specifications
Specifications
1.5 mm
Yes
50/Pk.
Borosilicate Glass (3.3)
10.0 NS