Learn More
DWK Life Sciences DURAN™ Standard Cone, finished
Product Code. 10181601 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
4 mm
6 mm
8 mm
11 mm
13 mm
17 mm
22 mm
26 mm
30 mm
36 mm
40 mm
46 mm
50 mm
56 mm
65 mm
80 mm
90 mm
Unit Size:
Each
Pack of 10
Pack of 50
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
10181601 90 mm Each
10706941 26 mm Each
15951531 13 mm Pack of 50
15864578 56 mm Pack of 10
15804568 17 mm Pack of 50
15844558 6 mm Pack of 10
15854558 11 mm Pack of 50
15804578 30 mm Pack of 50
15941531 8 mm Pack of 50
15961531 22 mm Pack of 50
15834578 40 mm Pack of 10
15844578 36 mm Pack of 10
15854578 46 mm Pack of 10
10191981 50 mm Pack of 10
15874578 65 mm Pack of 10
10305211 80 mm Each
15834558 4 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10181601 Supplier DWK Life Sciences Supplier No. 286160404
Only null left
Add to basket
Add to basket
For a rigid, self-supporting connection.
The conical ground joint is the most common joint in glass apparatus engineering. Ground DURAN™ cores comply with DIN 12249 Form B in their design and meet higher requirements for surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • According to DIN 12249 Form B
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts.
  • Minimized grease requirement
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Item Description Installation in glass apparatus and devices
Joint Size 90 mm
Thickness (Metric) Wall 3.5 mm