Learn More
DWK Life Sciences DURAN™ Spare mixer screw cap GLS 80™ only
Product Code. 10083534 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GLS 80
Unit Size:
Each
Product Code. Thread Size (Metric) unitSize
10083534 GLS 80 Each
1 options
Product Code. 10083534 Supplier DWK Life Sciences Supplier No. 1200385
Only null left
Add to basket
Add to basket
Easy to use GLS 80™ lab bottle based reactor system with tubing connectors for use with magnetic stirrers.
The DURAN™ stirred reactor is ideal for the widest possible range of mixing processes in laboratories, e. g. mixing liquids or dissolving solids. The connections available permit you to introduce other media into the bottle or remove media during mixing. The connections (2 x GL 14 and 2 x GL 18) provided permit addition or removal of media from the bottle during the mixing process. The whole unit can be autoclaved and is therefore suitable for biological applications. By using components from the GLS 80™ connection system, an additional media bottle (Tubing outer diameter: 1.6 - 12.0 mm) can be connected or a sterile pressure equalizer attached. Drive for the stirrer may provided by a standard commercial magnetic stirrer. The variable stirrer shaft can be used in DURAN™ GLS 80™ laboratory glass bottles (1000 mL and 2000 mL) and provides notably improved mixing in comparison with standard magnetic stir bars. The stirrer unit is interchangeable and can be used up to 500 rpm is possible
  • The variable stirrer shaft can be adjusted to the optimal position in either 500, 1000 or 2000 ml DURAN™ GL 45 bottles
  • Semi-rigid tubing with 1.6 to 6.0 mm bores can be used to add or remove liquid or gas
  • The connections (2 x GL 14 and 2 x GL 18) provided permit addition or removal of media from the bottle during the mixing process
  • Drive with standard magnetic stirrers up to 500 rpm and allows improved mixing compared to stir bars
  • Fully compatible with the proven DURAN™ connection systems components
TRUSTED_SUSTAINABILITY
Specifications
Description Spare screw cap for GLS 80 stirred reactor, PP, blue/grey
For Use With (Application) Simple, low cost, bioreactor; Simple continuous flow stirred reactor; Mixing liquids; Photo-bioreactor
Material Polypropylene, PTFE, PEEK, Stainless Steel
Quantity 1 Ea.
Thread Size (Metric) GLS 80
Neck Finish 20 mm
Autoclavable Autoclavable