Learn More
DWK Life Sciences DURAN™ Petri Dish, pressed
Product Code. 10142101 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
100 mm
100 x 15 mm
100 x 20 mm
Type:
Four-Sectional
Half-Sectional
Three-Sectional
Without Sections
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Type unitSize
10142101 100 mm Three-Sectional Pack of 10
11726958 100 mm Four-Sectional Pack of 10
10132101 100 mm Half-Sectional Pack of 10
10796771 100 x 20 mm Without Sections Pack of 10
15585016 100 x 15 mm Without Sections Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10142101 Supplier DWK Life Sciences Supplier No. 217534802
Only null left
Add to basket
Add to basket
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow. DURAN™ Petri dishes are manufactured from borosilicate glass 3.3 using a special manufacturing process. They allow an even distribution of the culture medium and guarantee a distortion-free view. The lids of the DURAN™ Petri dishes rest directly on the lower part and thus prevent turbulence of the protruding air space. This is well suited for reactions with long incubation times, at high temperatures or with sensitive culture media.
  • Facilitated fractionated dilution smear by division (into 2, 3 or 4 parts)
  • Available half-, three- and four-sectional or without sections
  • Pressed
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (3.3)
Type Three-Sectional
For Use With (Application) Cultivation on different culture media at the same time under the same conditions; Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; cell culture
No. per Pack 10/Pk.
Diameter (Metric) 100 mm