Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Product Code. 15201729 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
180 mL
220 mL
280 mL
380 mL
420 mL
450 mL
530 mL
770 mL
1250 mL
1650 mL
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15201729 180 mL Pack of 10
11746998 380 mL Pack of 10
15804508 220 mL Pack of 10
15231729 280 mL Pack of 10
15211729 420 mL Pack of 10
15221729 530 mL Pack of 10
15241729 450 mL Pack of 10
15251729 1650 mL Pack of 10
10334261 770 mL Each
15261729 1250 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15201729 Supplier DWK Life Sciences Supplier No. 213983406
Only null left
Add to basket
Add to basket
Cylinders without graduation can be used to mix components.
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
  • Rough ground rim
  • With round base
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Diameter (Metric) Outside 40 mm
For Use With (Application) Diluting solutions; Mixing several components
Capacity (Metric) 180 mL
Autoclavable Autoclavable
Quantity 10/Pk.