Learn More
DWK Life Sciences DURAN™ Micro Immersion Filter, for reverse filtration
The micro immersion filter is suitable for the extraction of supernatant solutions in very small quantities (clarification filtration).
Brand: DWK Life Sciences 258571102
Code : NEW
Additional Details : Weight : 0.05000kg
Description
Filtrations are possible in many different ways. A filtration variant at reduced pressure is clarification filtration, in which larger quantities of liquid are to be filtered. In this method, the precipitate is allowed to settle and the liquid above is sucked off through an immersion filter. This prevents the formation of a filter cake which could clog the filter and reduce the filtration speed.
- For reverse filtration
- Extraction of clear filtrate (not the filtrand)
- Fired-on, highly durable white print
Specifications
Porosity 1 | |
Borosilicate Glass (3.3) | |
Clarification filtration; Extraction of clear filtrate (not the filtrand); Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering | |
10/Pk. |