Learn More
DWK Life Sciences DURAN™ Micro Immersion Filter, for reverse filtration
The micro immersion filter is suitable for the extraction of supernatant solutions in very small quantities (clarification filtration).
Brand:  DWK Life Sciences 258571205
Code : A0
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 11810710
  • £281.00 / Pack of 10
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
Filtrations are possible in many different ways. A filtration variant at reduced pressure is clarification filtration, in which larger quantities of liquid are to be filtered. In this method, the precipitate is allowed to settle and the liquid above is sucked off through an immersion filter. This prevents the formation of a filter cake which could clog the filter and reduce the filtration speed.
  • For reverse filtration
  • Extraction of clear filtrate (not the filtrand)
  • Fired-on, highly durable white print
Specifications
Specifications
Porosity 2
Borosilicate Glass (3.3)
Clarification filtration; Extraction of clear filtrate (not the filtrand); Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering
10/Pk.