Learn More
DURAN™ Laboratory Glass Ceramic Lab Protection Plate
Due to low thermal expansion stresses, these glass ceramic plates are well suited to heating glassware with a Bunsen burner.
Brand:  Duran 238215806
Product Code. 10251171