Learn More
Lenz Laborglasinstrumente™ Duran Glass Straight Gas Inlet Tubes
Click to view available options
Length (Metric):
200mm
Unit Size:
Each
Specifications
Specifications
| Includes | NS cone and thin capillary |
| Length (Metric) | 200mm |
| Type | Gas Inlet Tube |
| Material | Duran Glass |
| Quantity | 1Each |
| Standard Taper Joint | NS 14/23 |