Learn More
Witeg™ Duran Glass Separatory Funnels
Product Code. 10750415
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
1000 mL
2000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10750415 100 mL Each
10632563 1000 mL Each
10081122 250 mL Each
10421763 50 mL Each
10787534 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10750415 Supplier Witeg™ Supplier No. 2 677 010
Witeg™ Duran™ Glass Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
  • Conical shape
  • Graduated
  • Economy-needle valve stopcock
  • PTFE-needle valve
  • Screw-thread and cap
Specifications
Capacity (Metric) 100 mL
Material Duran Glass, Borosilicate Glass
Quantity 1Pack
Product Type Separatory Funnel