Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
A funnel enables a clean transfer and filtration of liquids.
Brand:  DWK Life Sciences 213515708
Code : A0
Additional Details : Weight : 2.10000kg
 View more versions of this product
Product Code. 11702362
  • £344.00 / Pack of 10
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • With short stem
  • Due to its shape, it is suitable for liquid-solid filtration using a filter paper
  • Fired-on, highly durable white print
Specifications
Specifications
150 mm
Yes
10/Pk.
Boro 3.3
Decanting and filtration of liquids; The spillage of substances is avoided; Filtering