Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Expansion from sleeve to core.
Brand: DWK Life Sciences 286542601
Code : NEW
Additional Details : Weight : 0.01800kg
Description
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
- Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
Specifications
10/Pk. | |
Connection of two components with different standard ground joint sizes | |
Socket NS 24/29, Cone NS 29/32 |