Learn More
DWK Life Sciences DURAN™ Desiccator Lid, with knob, suitable for all types of bases
The desiccator lid is used to cover or seal the desiccator base.
Brand:  DWK Life Sciences 244105706
105.27 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Code : A0
Additional Details : Weight : 0.65500kg
 View more versions of this product
Product Code. 10712732
  • £114.00 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
The DURAN™ desiccator is a vacuum-suitable laboratory vessel with a ground lid, which is used for storing moisture-sensitive substances and for drying moist products. Separated by a sieve plate (made of metal or porcelain), a drying agent is introduced into the lower part of the desiccator base, which binds the moisture of the substance to be dried, placed on the sieve plate. To accelerate the drying process, a vacuum can be connected if necessary, as the liquid evaporates faster due to a lower pressure in the desiccator. The vacuum is applied via a shut-off valve connection. To facilitate loosening of the lid, the surface grinding of the desiccator must be well greased, since after evacuation the lid is pressed onto the lower part. All DURAN™ desiccators are vacuum-resistant, i.e. they can be used up to the technically maximum possible vacuum.
  • The lid can be opened easily while using the button
  • Suitable for all types of bases
Specifications
Specifications
DN 150
Borosilicate Glass 3.3
Ideal for removing moisture from solids; Storage of moisture-sensitive materials over long periods; For applications requiring vacuum desiccation; Drying of materials; Safe working under vacuum