missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Borosilicate Round Bottom Test Tubes

Product Code. 10756572
Click to view available options
Length (Metric):
75 mm
Diameter (Metric):
12 mm
Rimmed:
Yes
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10756572

Brand: Karl Hecht™ 42788024

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for routine laboratory analysis and procedures. Karl Hecht™ Assistent™ Borosilicate Round Bottom Test Tubes are manufactured from durable DURAN™ 3.3 borosilicate glass.

  • With or without rim
  • With heavy wall
  • Corrosion-free and resistant to thermal shock

Specifications

Design Type Round bottom, heavy wall
Length (Metric) 75 mm
Packaging 1Pack
Rimmed Yes
Diameter (Metric) 12 mm
Product Type Test Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.