Learn More
Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper
Product Code. 10339061
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
Closure Size:
NS 14/23
NS 19/26
NS 29/32
Material:
Borosilicate Glass (3.3)
Borosilicate Glass, Duran Glass
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Material unitSize
10339061 250 mL NS 29/32 Borosilicate Glass, Duran Glass Each
10778652 100 mL NS 19/26 Borosilicate Glass, Duran Glass Each
10399441 50 mL NS 29/32 Borosilicate Glass (3.3) Each
10043631 100 mL NS 19/26 Borosilicate Glass (3.3) Each
10728662 500 mL NS 29/32 Borosilicate Glass (3.3) Each
10207481 25 mL NS 14/23 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10339061 Supplier Karl Hecht™ Supplier No. 41918030
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper is constructed of durable Duran™ borosilicate 3.3 glass.
  • Durable Duran™ borosilicate 3.3 glass.
  • Graduated.
  • Reinforced ground neck marked with standard ground size.
  • Hollow NS glass stopper.
Specifications
Closure Material Glass
Capacity (Metric) 250 mL
Material Borosilicate Glass, Duran Glass
Includes Hollow glass stopper
Standard Taper Joint NS 29/32
Graduated Yes
Closure Type NS Stopper
Closure Size NS 29/32