Description
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
- Tight closure due to standard ground joint
Specifications
Specifications
| Quantity | 1 Ea. |
| Item Description | Connection of two glass components |
| Type | Expansion Joint |
| Joint Size | Socket NS 14/23, Cone NS 19/26 |