Learn More
Lenz Laborglasinstrumente™ Two-Neck Adapters
Provides for safe and secure connections to apparatus in the laboratory. Lenz Laborglasinstrumente™ Two-Neck Adapters are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05010044
Code : A0
Additional Details : Weight : 0.09900kg
Description
- Acc. to DIN 12594
- Two NS sockets and one NS cone
- High taper accuracy of the cone
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
Specifications
2-neck Adapter | |
Duran | |
With two parallel necks, with two NS sockets and one NS cone |