missing translation for 'onlineSavingsMsg'
Learn More

Duramycin (~90%), TRC

Product Code. 30380748
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380748 1 mg -
30376922 2.5 mg 2.5mg
30372489 0.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380748 Supplier Toronto Research Chemicals Supplier No. TRCD7350231MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391-36-2
Molecular Formula C89H125N23O25S3
Molecular Weight (g/mol) 2013.28
Synonym 27,46-(Methaniminopentaniminomethano)-6,43:15,40-bis(methanothiomethano)pyrrolo[2,1-d1][1,4,7,10,13,16,19,22,25,28,31,34,37,40,43,46,49,52]thiaheptadecaazacyclopentapentacontine, cyclic peptide deriv., Antibiotic PA 48009, Lancovutide, Leucopeptin, Moli 1901
SMILES CC(C)[C@@H]1NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@@H]3NC(=O)[C@H](Cc4ccccc4)NC(=O)[C@@H]5CCCN5C(=O)CNC(=O)[C@H](Cc6ccccc6)NC(=O)[C@@H]7CNCCCC[C@H](NC(=O)[C@@H]8NC(=O)[C@H](CC(=O)N)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CSC[C@@H](NC(=O)[C@H](CCC(=O)N)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CS[C@H]8C)C(=O)N[C@@H](CS[C@H]3C)C(=O)N7)NC1=O)[C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C89H125N23O25S3/c1-44(2)67-85(131)107-60-42-138-41-59-80(126)106-61-43-140-46(4)69(87(133)103-56(76(122)108-67)35-49-23-12-7-13-24-49)109-77(123)55(34-48-21-10-6-11-22-48)102-83(129)62-27-18-32-112(62)66(116)39-96-73(119)54(33-47-19-8-5-9-20-47)101-79(125)58(104-81(61)127)37-94-31-17-15-26-53(88(134)135)100-86(132)68(110-78(124)57(36-64(93)114)97-65(115)38-95-84(130)70(111-82(60)128)71(117)89(136)137)45(3)139-40-50(91)72(118)98-51(25-14-16-30-90)74(120)99-52(75(121)105-59)28-29-63(92)113/h5-13,19-24,44-46,50-62,67-71,94,117H,14-18,25-43,90-91H2,1-4H3,(H2,92,113)(H2,93,114)(H,95,130)(H,96,119)(H,97,115)(H,98,118)(H,99,120)(H,100,132)(H,101,125)(H,102,129)(H,103,133)(H,104,127)(H,105,121)(H,106,126)(H,107,131)(H,108,122)(H,109,123)(H,110,124)(H,111,128)(H,134,135)(H,136,137)/t45-,46-,50-,51-,52-,53-,54-,55-,56-,57-,58-,59+,60-,61-,62-,67-,68+,69+,70-,71+/m0/s1
Formula Weight 2011.8379
Chemical Name or Material Duramycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.