Learn More
DuraCal™ 7.00 Value pH Buffer, Hamilton™
Product Code. 10199151
Change view
Click to view available options
Quantity:
3 x 500 mL
Unit Size:
Pack of 3
Product Code. Quantity unitSize
10199151 3 x 500 mL Pack of 3
1 options
Product Code. 10199151 Supplier Hamilton Supplier No. 238918
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 7.00 Value pH Buffers have actual values determined by a DKD laboratory, accreditd for pH measurement.
  • 7.00 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
pH 7.00
Quantity 3 x 500 mL