missing translation for 'onlineSavingsMsg'
Learn More

DuraCal™ Buffer pH 10.01, Hamilton™

Product Code. 10069134
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
1500mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10069134 250 mL 250mL
10332982 500 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10069134 Supplier Hamilton Supplier No. 238321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 10.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.

  • 10.01 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards

Specifications

Color Yellow
pH 10.01
Physical Form Liquid
Quantity 250 mL
Solution Type Buffer
Solubility Information Water Soluble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.