Learn More
DuPont™ Tyvek™ IsoClean™ Sleeves
Product Code. 11783113
Product Code. 11783113 Supplier DuPont™ Supplier No. D14238500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Sterile sleeve available in white and in one size
  • Tunnelled elastication at wrist and upper arm
  • This accessory is not CE certified as chemical protective clothing
  • Gamma-sterilised
  • Sterility assurance level (SAL) of 10-6
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
  • Full traceability on all sterilized apparel with certificates of sterility available
  • Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
  • Tyvek™ is permeable to both air and water vapour, yet repels water-based liquids and aerosols
TRUSTED_SUSTAINABILITY
Specifications
Model D14238500