missing translation for 'onlineSavingsMsg'
Learn More

Duplex 2209, 20 mesh woven from 0.3mm (0.012mm) dia wire, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 046416.GT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15477205

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Duplex 2209
150 x 150 mm
Odorless
20 mesh woven from 0.3mm (0.012mm) dia. wire
20 Mesh
Wire Cloth
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers