missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUOXA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26632-1-AP-150UL
This item is not returnable.
View return policy
Description
Dual oxidases DUOX1 and DUOX2 are NADPH oxidases which are involved in hydrogen peroxide production necessary for thyroid hormonogenesis. They form a heterodimer with specific maturation factors DUOXA1 and DUOXA2, respectively, which is essential for the maturation and function of the DUOX enzyme complexes. This gene encodes the DUOX1 activator or maturation factor DUOXA1. Rat studies identified a bidirectional promoter which controls the transcription of the DUOX1 and DUOXA1 genes. This protein is cotransported to the cell surface when coexpressed with DUOX1 and is retained in the endoplasmic reticulum when expressed without DUOX1 protein. The expression of this gene or the DUOX1 gene is not suppressed by thyroglobulin (Tg), a macromolecular precursor in thyroid hormone synthesis, while the expression of the DUOX2 and DUOXA2 are significantly suppressed by the Tg. This protein is also a p53-regulated neurogenic factor involved in p53 dependent neuronal differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| DUOXA1 | |
| Polyclonal | |
| Unconjugated | |
| DUOXA1 | |
| Dual oxidase activator 1, DUOXA1, mol, NIP, Numb interacting protein, NUMBIP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 213696, 90527 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q1HG43, Q8VE49 | |
| DUOXA1 | |
| DUOXA1 Fusion Protein Ag24373 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction