missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUOX1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142802
This item is not returnable.
View return policy
Description
Additional Information: This antibody is not expected to cross-react with DUOX2. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX2. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
Specifications
| DUOX1 | |
| Polyclonal | |
| Unconjugated | |
| DUOX1 | |
| 9930101G15Rik; AW987690; Dual oxidase 1; dual oxidase 2; DUOX; DUOX1; Duox2; flavoprotein NADPH oxidase; large NOX 1; LNOX1; LNOX2; long NOX 1; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF1; NOXEF2; P138-TOX; THOX1; THOX2; thyroid oxidase 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 53905 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9NRD9 | |
| DUOX1 | |
| Peptide with sequence CSRSDIETELTPQR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction