missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DUOX1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585452

Product Code. 16392385

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (89%), Rat (87%), Dog (86%), Pig (90%), Bovine (90%). Positive Control: A549, H1299, HCT116, unboiled A431, A431 membrane extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX2. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DUOX1
Polyclonal
Unconjugated
DUOX1
9930101G15Rik; AW987690; Dual oxidase 1; dual oxidase 2; DUOX; DUOX1; Duox2; flavoprotein NADPH oxidase; large NOX 1; LNOX1; LNOX2; long NOX 1; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF1; NOXEF2; P138-TOX; THOX1; THOX2; thyroid oxidase 1
Rabbit
Antigen affinity chromatography
RUO
53905
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q9NRD9
DUOX1
Recombinant protein encompassing a sequence within the Extracellular domain of human DUOX1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.