missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £470.00
Specifications
| Antigen | DUOX1 |
|---|---|
| Concentration | 0.05mg/mL |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458070
|
Novus Biologicals
NBP1-81301-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284787
|
Novus Biologicals
NBP1-81301 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DUOX1 Polyclonal specifically detects DUOX1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DUOX1 | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| dual oxidase 1, DUOX, EC 1.11.1.-, EC 1.6.3.1, Large NOX 1, LNOX1flavoprotein NADPH oxidase, Long NOX 1, MGC138840, NADPH thyroid oxidase 1, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF1, THOX1MGC138841, Thyroid oxidase 1 | |
| DUOX1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.05mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 53905 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QHKEELTWEDFHFMLRDHNSELRFTQLCVKGVEVPEVIKDLCRRASYISQDMICPSPRVSARCSRSDIETELTPQRLQCPMDTDPPQEIRRRFGKKVTSFQP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title