missing translation for 'onlineSavingsMsg'
Learn More

Duocarmycin TM, MedChemExpress

Product Code. 16494148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494148 10 mg 10mg
16476857 5 mg 5mg
16445116 100 mg 100mg
16412976 25 mg 25mg
16400507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494148 Supplier Medchem Express Supplier No. HY107769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Duocarmycin TM (CBI-TMI) is a potent antitumor antibiotic. Duocarmycin TM induces a sequence-selective alkylation of duplex DNA.

Duocarmycin TM (CBI-TMI) is a potent antitumor antibiotic. Duocarmycin TM induces a sequence-selective alkylation of duplex DNA.

Specifications

Chemical Name or Material Duocarmycin TM
Physical Form Solid
CAS 157922-77-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H23ClN2O5
For Use With (Application) Cancer-programmed cell death
Synonym CBI-TMI
Solubility Information DMSO : ≥ 50 mg/mL (107.09 mM)
SMILES ClC[C@H]1C2=C(C=CC=C3)C3=C(O)C=C2N(C(C4=CC(C=C(OC)C(OC)=C5OC)=C5N4)=O)C1
Molecular Weight (g/mol) 466.91
Formula Weight 466.91
Percent Purity 95.04%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.