missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duloxetine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Duloxetine hydrochloride |
| CAS | 136434-34-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H19 N O S . Cl H |
| Quantity | 1 g |
| Synonym | 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride (1:1), (γS)-, 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride, (S)-, 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride, (γS)- (9CI), (S)-(+)-Duloxetine hydrochloride, (S)-(+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, (S)-Duloxetine hydrochloride, (S)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine hydrochloride, (S)-γ-(1-Naphthalenoxy)-N-methylthiophene-2-propanamine monohydrochloride, Cymbalta, Dulane 20, Duloxetine hydrochloride, LY 248686 HCl, (γS)-N-Methyl-γ-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride |
| InChI Formula | InChI=1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m0./s1 |
| SMILES | Cl.CNCC[C@H](Oc1cccc2ccccc12)c3cccs3 |
| IUPAC Name | (3S)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-2-ylpropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 333.88 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?