missing translation for 'onlineSavingsMsg'
Learn More

Dulcin, Dr. Ehrenstorfer

Product Code. 30326285
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326285 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326285 Supplier Dr. Ehrenstorfer Supplier No. DREC13097500
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycosine (Dulcin)
CAS 150-69-6
Analyte or Component Names Glycosine (Dulcin)
Shipping Condition +20°C
Molecular Formula C9 H12 N2 O2
Certifications/Compliance ISO/IEC 17025
Synonym Urea, (4-ethoxyphenyl)- (9CI), Urea, (p-ethoxyphenyl)- (8CI), N-(4-Ethoxyphenyl)urea, (4-Ethoxyphenyl)urea, (p-Ethoxyphenyl)urea, 1-(4-Ethoxyphenyl)urea, 4-Ureidophenetole, Dulcin, Dulcin (sweetener), Dulcine, NSC 1839, Phenetolcarbamide, Sucrol, Valzin, p-Phenetolcarbamide, p-Phenetylurea
InChI Formula InChI=1S/C9H12N2O2/c1-2-13-8-5-3-7(4-6-8)11-9(10)12/h3-6H,2H2,1H3,(H3,10,11,12)
SMILES O=C(N)NC1=CC=C(OCC)C=C1
IUPAC Name (4-ethoxyphenyl)urea
Molecular Weight (g/mol) 180.2
Quantity 25 mg
Formula Weight 180.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.